missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Cystatin E/M ELISA Kit
Klik for at se tilgængelige muligheder
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Pakningsstørrelse:
Each
Pack of 10
Pack of 96
Beskrivelse
Human Cystatin E/M quantitates human Cystatin E/M in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Cystatin E/M.
The cystatin superfamily is a well-established family of cysteine protease inhibitors. Cystatins A and B (type 1) are mainly intracellular; cystatins C, D, E/M, F, G, S, SN and SA cystatins are extracellular (type 2); and the kininogens are type 3 cystatins which are intravascular proteins. All true cystatins inhibit cysteine peptidases of the papain family, such as cathepsins, and some also inhibit legumain family enzymes. Cystatin SA, cystatin S and cystatin SN are found primarily in saliva. Cystatin S and SN can also be expressed in tears, urine and seminal fluid. Cystatin C is a related protein which is expressed in brain, thymus, ovary, epididymis and vas deferens. Cystatin D protects against proteinases in the oral cavity, while Cystatin E/M and F moderate the inhibition of cathepsin proteins. The fetuins, part of the cystatin superfamily, are secretable proteins that influence osteogenesis and bone resorption, regulation of the insulin and hepatocyte growth factor receptors and the response to systemic inflammation. High molecular weight kininogen (Kininogen HC) and low molecular weight kininogen (Kininogen LC) have varied roles, though they both inhibit the thrombin- and plasmin-induced aggregation of thrombocytes.
Tekniske data
Tekniske data
| Accession Number | 1110017E11Rik; CST6; cystatin 6; cystatin E/M; cystatin M; cystatin M/E; cystatin N; cystatin-6; Cystatin-E; Cystatin-M; cysteine proteinase inhibitor; harlequin ichthyosis; harlequin ichthysosis; ichq; N28197 |
| Assay Range | 0.14 to 30 ng/mL |
| Assay Sensitivity | 0.14 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 1474 |
| Gene Symbol | CST6 |
| Interassay CV | <12% |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion