missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cystatin C Antibody, R&D Systems™

Mouse Monoclonal Antibody
£158.00 - £524.00
Specifications
| Antigen | Cystatin C |
|---|---|
| Clone | 197814 |
| Dilution | Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18724127
![]() |
R&D Systems
MAB11963-SP |
25 μg |
£158.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18701497
![]() |
R&D Systems
MAB11963-100 |
100 μg |
£524.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cystatin C Monoclonal antibody specifically detects Cystatin C in Human samples. It is validated for Immunohistochemistry, ImmunocytochemistrySpecifications
| Cystatin C | |
| Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| ARMD11, bA218C14.4 (cystatin C), CST3, cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, Post-gamma-globulin | |
| Mouse myeloma cell line NS0-derived recombinant human Cystatin C, Ser27-Ala146, Accession # P01034 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 197814 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P01034 | |
| 1471 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title