missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cystatin C Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB11962-500
This item is not returnable.
View return policy
Description
Cystatin C Monoclonal specifically detects Cystatin C in Human samples. It is validated for Immunohistochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| Cystatin C | |
| Monoclonal | |
| LYOPH | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P01034 | |
| CST3 | |
| Mouse myeloma cell line NS0-derived recombinant human Cystatin C Ser27-Ala146 Accession # P01034 | |
| 500 μg | |
| Primary | |
| Detects human Cystatin C in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human Cystatins A, B, D, E/M, F, S, SA, SN, X/Z/P, or recombinant mouse Cystatin C is observed. | |
| Human | |
| Supernatant |
| ELISA | |
| 197820 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARMD11, bA218C14.4 (cystatin C), CST3, cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, Post-gamma-globulin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1471 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction