missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Cyr61/CCN1 Quantikine ELISA Kit

Product Code. 18731964
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For unitSize
18731964 1 x 96-well Plate 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18731964 Supplier R&D Systems™ Supplier No. DCYR10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Cyr61/CCN1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Cyr61/CCN1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Cyr61/CCN1 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), saliva (50 ul), human milk (10 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 39.10 - 2500pg/mL
Assay Sensitivity 3.8pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Human Milk, Plasma (EDTA), Saliva, Serum
Specificity Natural and Recombinant Human Cyr61
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 3491
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Saliva (50uL),Human Milk (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CCN family member 1, CCN1, CCN1IGF-binding protein 10, Cysteine-rich angiogenic inducer 61, cysteine-rich, angiogenic inducer, 61, GIG1, GIG1cysteine-rich heparin-binding protein 61, IBP-10, IGFBP-10, IGFBP10cysteine-rich, anigogenic inducer, 61, Insulin-like growth factor-binding protein 10, protein CYR61, Protein GIG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.