missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CYP7A1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9120-100
This item is not returnable.
View return policy
Description
CYP7A1 Monoclonal specifically detects CYP7A1 in Human samples. It is validated for Immunocytochemistry.
Specifications
| CYP7A1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cholesterol 7-alpha-hydroxylase, cholesterol 7-alpha-monooxygenase, CP7A, CYP7, CYP7MGC126826, CYPVII, Cytochrome P450 7A1, cytochrome P450, family 7, subfamily A, polypeptide 1, cytochrome P450, subfamily VIIA (cholesterol 7 alpha-monooxygenase), cytochrome P450, subfamily VIIA polypeptide 1, EC 1.14.13, EC 1.14.13.17, MGC138389, polypeptide 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1581 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Immunocytochemistry | |
| 946902 | |
| Immunocytochemistry 1-25 ug/mL | |
| P22680 | |
| CYP7A1 | |
| Human CYP7A1 synthetic peptide Accession # P22680 | |
| 100 μg | |
| Primary | |
| Detects human CYP7A1 in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion