Learn More
Abnova™ Human CYP4F22 Full-length ORF (AAH69351.1, 1 a.a. - 531 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This gene is part of a cluster of cytochrome P450 genes on chromosome 19 and encodes an enzyme thought to play a role in the 12(R)-lipoxygenase pathway. Mutations in this gene are the cause of ichthyosis lamellar type 3. [provided by RefSeq]
Specifications
Specifications
| Accession Number | AAH69351.1 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 126410 |
| Molecular Weight (g/mol) | 88.4kDa |
| Name | CYP4F22 (Human) Recombinant Protein (P01) |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 10 μg |
| Immunogen | MLPITDRLLHLLGLEKTAFRIYAVSTLLLFLLFFLFRLLLRFLRLCRSFYITCRRLRCFPQPPRRNWLLGHLGMYLPNEAGLQDEKKVLDNMHHVLLVWMGPVLPLLVLVHPDYIKPLLGASAAIAPKDDLFYGFLKPWLGDGLLLSKGDKWSRHRRLLTPAFHFDILKPYMKIFNQSADIMHAKWRHLAEGSAVSLDMFEHISLMTLDSLQKCVFSYNSNCQEKMSDYISAIIELSALSVRRQYRLHHYLDFIYYRSADGRRFRQACDMVHHFTTEVIQERRRALRQQGAEAWLKAKQGKTLDFIDVLLLARDEDGKELSDEDIRAEADTFMFEGHDTTSSGISWMLFNLAKYPEYQEKCREEIQEVMKGRELEELEWDDLTQLPFTTMCIKESLRQYPPVTLVSRQCTEDIKLPDGRIIPKGIICLVSIYGTHHNPTVWPDSKVYNPYRFDPDNPQQRSPLAYVPFSAGPRNCIGQSFAMAELRVVVALTLLRFRLSVDRTRKVRRKPELILRTENGLWLKVEPLPPRA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.