missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CYP4B1 (aa 64-186) Control Fragment Recombinant Protein

Product Code. 30200479
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200479

Brand: Invitrogen™ RP101024

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82148 (PA5-82148. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum. In rodents, the homologous protein has been shown to metabolize certain carcinogens; however, the specific function of the human protein has not been determined. Two transcript variants encoding slightly different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13584
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 1580
Name Human CYP4B1 (aa 64-186) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CYP4B1; Cyp4b-1; CYP4B1 isoform; CYP4B1-like isozyme short form; CYPIVB1; cytochrome P450 4B1; cytochrome P450 family 4 subfamily B member 1; cytochrome P450 isozyme 5; cytochrome P-450 isozyme 5; Cytochrome P450 L-2; cytochrome P450, family 4, subfamily B, polypeptide 1; cytochrome P450, subfamily 4 B, polypeptide 1; cytochrome P450, subfamily IV B, polypeptide 1; cytochrome P450, subfamily IVB, polypeptide 1; cytochrome P450-HP; microsomal monooxygenase; P450 L-2; P-450 HP; P450-isozyme 5
Common Name CYP4B1
Gene Symbol CYP4B1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TGSLDKVVSWAHQFPYAHPLWFGQFIGFLNIYEPDYAKAVYSRGDPKAPDVYDFFLQWIGRGLLVLEGPKWLQHRKLLTPGFHYDVLKPYVAVFTESTRIMLDKWEEKAREGKSFDIFCDVGH
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.