missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CYP26B1 (aa 201-310) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52948 (PA5-52948. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and the synthesis of cholesterol, steroids and other lipids. The enzyme encoded by this gene is involved in the specific inactivation of all-trans-retinoic acid to hydroxylated forms, such as 4-oxo-, 4-OH-, and 18-OH-all-trans-retinoic acid.
Specifications
Specifications
| Accession Number | Q9NR63 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 56603 |
| Name | Human CYP26B1 (aa 201-310) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | CP26; CYP26A2; CYP26B1; Cytochrome P450 26A2; Cytochrome P450 26B1; cytochrome P450 family 26 subfamily A member 1; cytochrome P450 family 26 subfamily B member 1; cytochrome P450 retinoic acid-inactivating 2; cytochrome P450 retinoid metabolizing protein; cytochrome P450, 26, retinoic acid B1; cytochrome P450, family 26, subfamily b, polypeptide 1; cytochrome P450, subfamily XXVIB, polypeptide 1; cytochrome P450RAI-2; dol; fc21d03; P450 26A2; P450 enzyme to degrade retinoic acid; P450RAI2; P450RAI-2; retinoic acid B1; retinoic acid hydroxylase; retinoic acid-metabolizing cytochrome; RHFCA; stocksteif; wu:fc21d03; wu:fc26h10; zgc:76999 |
| Common Name | CYP26B1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction