missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human/Cynomolgus Monkey PD-L1/B7-H1 QuicKit ELISA
Specifications
Specifications
| Assay Range | 14.10 to 900 pg/mL |
| Assay Sensitivity | 2.29 pg/mL |
| Sample Type | Cell Culture Supernatant, Cell Lysates, Plasma (EDTA), Serum, Plasma (Heparin) |
| Specificity | Natural and recombinant human PD-L1/B7-H1 and natural cynomolgus monkey PD-L1/B7-H1. |
| Target Species | Human, Cynomolgus Monkey |
| Gene ID (Entrez) | 29126 |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?