missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Cyclophilin A Recombinant Protein

Product Code. 11554920
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 11554920

Brand: Invitrogen™ RP020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

RP-020 can be used to catalyze the cis-trans-isomerization of X-Pro-peptide bonds. Protein folding reactions that are limited by the isomerization of X-Pro-peptide bonds are accelerated by cyclophilin A. RP-020 is a full-length human recombinant Cyclophilin A protein expressed in E. coli. Caution: Loss of protein may occur at high dilutions due to adsorption to container surfaces. Freezing of the enzyme or storage of the enzyme in diluted form could lead to loss of activity. In order to best maintain the endotoxin specification and peptidyl prolyl isomerase activity, it is recommended that aliquots be withdrawn with sterile pipet tips, if possible under a sterile hood.

Immunophilins are a family of soluble cytosolic receptors capable of binding to one of two major immunosuppressant agents - cyclosporin A (CsA) or FK506. Proteins that bind FK506 are termed FK506 Binding Proteins (FKBPs) and those that bind cyclosporin A are called cyclophilins (CyP). Both CyP:CsA and FKBP:FK506 complexes have been shown to inhibit calcineurin, a calcium and calmodulin dependent protein phosphatase which has been implicated as an important signaling enzyme in T-cell activation, providing a possible mechanism of immunosuppression by CsA and FK506. Immunophilins function as peptidyl prolyl cis-trans-isomerases (PPIase) whose activity is inhibited by their respective immunosuppressant compounds. As PPIase's, immunophilins accelerate folding of some proteins both in vivo and in vitro by catalyzing slow steps in the initial folding and rearrangement of proline containing proteins. Within the cyclophilin family, there are several different proteins which show a high degree of homology including CyPA, CyPB and CyPC. CyPA, also termed CyP-18, is the most abundant and ubiquitous cyclophilin found in all vertebrate tissues and is present in T-cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P62937
Concentration 0.8 mg/mL
For Use With (Application) Control, Binding Assay
Formulation 20mM tris HCl with no preservative; pH 7.8
Gene ID (Entrez) 5478
Molecular Weight (g/mol) 18 kDa
Name Human Cyclophilin A
pH Range 7.8
Purification Method Purified
Quantity 100 μg
Source E. coli
Storage Requirements 4°C
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Gene Alias 2700098C05; Cphn; CYCA; cyclophilin; Cyclophilin A; Cyclosporin A-binding protein; CyP A; CyP-18; CYPA; CyP-A; CYPH; EC 5.2.1.8; epididymis secretory sperm binding protein Li 69p; HEL-S-69p; HGNC:9253; I79_011869; MGC117158; MGC12404; MGC23397; p1B15; p31; Peptidyl-prolyl cis-trans isomerase A; Peptidyl-prolyl cis-trans isomerase A, N-terminally processed; peptidylprolyl isomerase A; peptidylprolyl isomerase A (cyclophilin A); PPIA; PPIase; PPIase A; rotamase; rotamase A; SP18; T cell cyclophilin
Common Name Cyclophilin A
Gene Symbol Ppia
Biological Activity Catalyzes the cis-trans-isomeration of X-Pro-peptide bonds. The protein folding that is limited by the isomeration of X-Pro-peptide bonds can be accelerated by cyclophilin A. Also binds to cyclosporins and the cyclophilin A/cyclosporin A.
Product Type Protein
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Sequence Full-length
Content And Storage 4°C
Activity Catalyzes the cis-trans-isomeration of X-Pro-peptide bonds. The protein folding that is limited by the isomeration of X-Pro-peptide bonds can be accelerated by cyclophilin A. Also binds to cyclosporins and the cyclophilin A/cyclosporin A.
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.