missing translation for 'onlineSavingsMsg'
Learn More

Human CXCR7/RDC-1 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£146.00 - £444.00

Specifications

Antigen CXCR7/RDC-1
Clone 358440
Dilution Immunohistochemistry 0.5-25 ug/mL, Immunocytochemistry 8-25 ug/mL
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18771337
GREENER_CHOICE
View Documents
R&D Systems
MAB42274-SP
25 μg
£146.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18741037
GREENER_CHOICE
View Documents
R&D Systems
MAB42274-100
100 μg
£444.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CXCR7/RDC-1 Monoclonal antibody specifically detects CXCR7/RDC-1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry
Specifications

Specifications

CXCR7/RDC-1
Immunohistochemistry 0.5-25 ug/mL, Immunocytochemistry 8-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
ACKR3, chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CMKOR1, CXCR7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog
NS0 mouse myeloma cell line transfected with human CXCR7/RDC-1, Met1-Lys362, Accession # AAA62370
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
358440
Immunohistochemistry, Immunocytochemistry
Unconjugated
Mouse
Human
AAA62370
57007
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.