missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCR4 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 13 publications
Brand: R&D Systems MAB171-100
This item is not returnable.
View return policy
Description
CXCR4 Monoclonal specifically detects CXCR4 in Human samples. It is validated for Neutralization.
Specifications
| CXCR4 | |
| Monoclonal | |
| LYOPH | |
| Neutralization 0.2-1.2 ug/mL | |
| CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201E, D2S201ESDF-1 receptor, FB22, Fusin, HM89, HM89NPYRL, HSY3RR, LAP3, LAP-3, LCR1, LESTR, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3R, NPY3RWHIM, NPYR, NPYRL, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor, WHIMS | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 7852 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Neutralization | |
| 44708 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CXCR4 | |
| 3T3 cells transfected with human CXCR4 | |
| 100 μg | |
| Primary | |
| Reacts specifically with human and non-human cells expressing human CXCR4 (fusin) as detected by flow cytometry. It will also react with cells expressing feline CXCR4 but not rat CXCR4. This antibody does not cross-react with other chemokine receptors. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction