missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CXCR2 (aa 3-48) Control Fragment Recombinant Protein

Product Code. 30209580
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209580

Brand: Invitrogen™ RP102608

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (46%), Rat (46%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83213 (PA5-83213. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CXCR2 is a receptor for interleukin-8, which is a powerful neutrophil chemotactic factor. It is a member of the GPCR family. Binding of IL-8 to the receptor causes activation of neutrophils. This response is mediated via a G-protein that activate a phosphatidylinositol-calcium second messenger system. This receptor binds to IL-8 with a high affinity and to GRO/MGSA and NAP-2 also with a high affinity. It has been reported to be expressed in a wide variety of tissues. ESTs have been isolated from human placenta and thymus libraries.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P25025
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3579
Name Human CXCR2 (aa 3-48) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; CD128; CD182; CD182 antigen; CDw128; CDw128b; chemokine (C-X-C motif) receptor 2; chemokine (CXC) receptor 2; chemokine (C-X-C) receptor 2; Cmkar2; CXC; C-X-C chemokine receptor type 2; CXC motif chemokine; C-X-C motif chemokine receptor 2; Cxcr2; CXC-R2; CXCR-2; CXCR2 gene for IL8 receptor type B; Gpcr16; GRO/MGSA receptor; high affinity interleukin-8 receptor B; IL-8 receptor alpha chain; Il-8 receptor b; IL-8 receptor beta; IL-8 receptor type 2; IL-8 R B; IL8R2; IL8RA; Il8rb; IL-8 rb; IL-8 Rh; interleukin 8 receptor B; interleukin 8 receptor type 2; interleukin 8 receptor, beta; interleukin-8 receptor type B; mIL-8 RH
Common Name CXCR2
Gene Symbol CXCR2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DFNMESDSFEDFWKGEDLSNYSYSSTLPPFLLDAAPCEPESLEINK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.