missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-8/CXCL8 DuoSet ELISA

Product Code. p-4754147
Click to view available options
Quantity:
1 Kit (for 15 Plates)
1 Kit (for 5 Plates)
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30021595

Brand: R&D Systems™ DY208

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Requires

DuoSet Ancillary Reagent Kit 2 - Catalog # DY008

OR

  • Wash Buffer - 0.05%Tween™ 20 in PBS, pH 7.2-7.4
  • Block Buffer - 1% BSA in PBS, pH 7.2-7.4
  • Reagent Diluent - 0.1% BSA, 0.05% Tween 20 in Tris-buffered Saline, pH 7.2-7.4, 0.2μm filtered
  • Substrate Solution - Catalog # DY999
  • Stop Solution - 2N H2SO4
  • Microplates - Catalog # DY990
  • ELISA Plate Sealers

Human IL-8/CXCL8 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-8/CXCL8 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human CXCL8/IL-8.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Solid Phase Sandwich ELISA Development Kit
Quantity 1 Kit (for 15 Plates)
Species Human
Conjugate Biotin
Assay Sensitivity 31.20 to 2000 pg/mL
Assay Range 31.20 to 2000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 3576
Synonym 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1
Kit Contents Capture Antibody
Detection Antibody
Recombinant Standard
Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.