missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCL7/NAP-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB393-SP
This item is not returnable.
View return policy
Description
CXCL7/NAP-2 Monoclonal specifically detects CXCL7/NAP-2 in Human samples. It is validated for Western Blot, Simple Western, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| CXCL7/NAP-2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Beta-TG, connective tissue-activating peptide III, CTAP III, CTAP3CTAP-III, CTAPIII, CXC chemokine ligand 7, C-X-C motif chemokine 7, CXCL7b-TG1, LA-PF4, LDGFTC1, Leukocyte-derived growth factor, low-affinity platelet factor IV, Macrophage-derived growth factor, MDGFTC2, NAP2, NAP-2, NAP-2-L1, neutrophil-activating peptide 2, neutrophil-activating peptide-2, PBPTHBGB, platelet basic protein, PPBP, pro-platelet basic protein (chemokine (C-X-C motif) ligand 7), SCAR10, SCYB7beta-thromboglobulin, small inducible cytokine B7, small inducible cytokine subfamily B, member 7, Small-inducible cytokine B7, TGB, TGB1B-TG1, THBGB1, thrombocidin 1, thrombocidin 2, thromboglobulin, beta-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5473 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Neutralization, ELISA | |
| 59418 | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Neutralization 15-20 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P02775 | |
| PPBP | |
| E. coli-derived recombinant human CXCL7/NAP-2 Ala59-Asp128 Accession # P02775 | |
| 25 μg | |
| Primary | |
| Detects human CXCL7/NAP-2 in ELISAs and Western blots. In sandwich ELISAs, less than 0.05% cross-reactivity with recombinant human (rh) CXCL1, 2, 3, 5, 6, 8, 9, 10, or 12 is observed. In Western blots, no cross-reactivity with rhCXCL1, 2, 3, 5, 6, 8, 9, 10, 12, 13, recombinant mouse CXCL1, 2, 6, 9, 10, 12 or recombinant rat CXCL1, 2 or 3 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction