missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCL4/PF4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
Brand: R&D Systems MAB7951-100
This item is not returnable.
View return policy
Description
CXCL4/PF4 Monoclonal specifically detects CXCL4/PF4 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| CXCL4/PF4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF4, PF-4, platelet factor 4, SCYB4oncostatin-A | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5196 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 170131 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P02776.2 | |
| PF4 | |
| E. coli-derived recombinant human CXCL4/PF4 Glu32-Ser101 Accession # P02776.2 | |
| 100 μg | |
| Primary | |
| Detects human CXCL4/PF4 in ELISAs. In ELISAs, this antibody does not cross-react with recombinant human (rh) CXCL1, rhCXCL2, or rmCXCL4. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction