missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCL4/PF4 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB795
This item is not returnable.
View return policy
Description
CXCL4/PF4 Monoclonal specifically detects CXCL4/PF4 in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| CXCL4/PF4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF4, PF-4, platelet factor 4, SCYB4oncostatin-A | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5196 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 170106 | |
| Western Blot 2 ug/mL, Simple Western 10 ug/mL | |
| P02776.2 | |
| PF4 | |
| E. coli-derived recombinant human CXCL4/PF4 Glu32-Ser101 Accession # P02776.2 | |
| 500 μg | |
| Primary | |
| Detects human CXCL4/PF4 in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse CXCL4/PF4, recombinant human (rh) GRO alpha , or rhGRO beta . | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction