missing translation for 'onlineSavingsMsg'
Learn More

Human CXCL17/VCC-1 Antibody, R&D Systems™

Product Code. 18199610
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18199610 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18199610 Supplier R&D Systems Supplier No. MAB4207

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

CXCL17/VCC-1 Monoclonal specifically detects CXCL17/VCC-1 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL17/VCC-1
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 422208
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q6UXB2
Gene Alias Chemokine (C-X-C Motif) Ligand 17, C-X-C Motif Chemokine 17, CXCL17, Dcip1, Dendritic Cell And Monocyte Chemokine-Like Protein, DMC, UNQ473, VCC1, VCC-1, VEGF Coregulated Chemokine 1, VEGF Co-Regulated Chemokine 1
Gene Symbols CXCL17
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL17/VCC-1 Leu24-Leu119 Accession # Q6UXB2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284340
Test Specificity Detects human CXCL17/VCC-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant mouse CXCL17 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.