missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CXCL12 (SDF-1a) Recombinant Protein

Recombinant Protein

Brand:  Invitrogen™ RP8658

Code : LE

Additional Details : Weight : 0.01000kg

Product Code. 11584960

  • £342.00 / 10µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CXCL12 (human) Recombinant Protein, RP-8658, from Invitrogen.

Stromal cell Derived Growth Factor-1 beta (SDF-1beta), also called CXCL12, is one of two splice variants made by a wide variety of cells when stimulated by inflammatory cytokines such as, TNF, IL-1 or LPS. SDF-1beta signals through the G protein-couple receptor, CXCR4, to recruit activated leukocytes. Human SDF-1beta shares 100% identity to feline SDF-1beta. Recombinant human SDF-1alpha is a non-glycosylated protein, containing 68 amino acids, with a molecular weight of 8 kDa. Purity is typically greater than 98% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the ability to chemoattract human peripheral T cells at 10-75 ng/mL. The specific activity of this protein is 1x10^5 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNK.
Specifications

Specifications

P48061
Control
6387
10μg
-20°C, Avoid Freeze/Thaw Cycles
H-SDF-1-alpha
CXCL12
Unconjugated
E. coli
Lyophilized
purified
0.1mg/mL
0.1%TFA with no preservative
Human CXCL12 alpha
E. coli
RUO
Human CXCL12 alpha
Human
Hu
Recombinant
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

in vitroresearch only