missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CXCL12/SDF-1 alpha Quantikine ELISA Kit

Product Code. 18772374
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18772374

Brand: R&D Systems™ DSA00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human CXCL12/SDF-1 alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human CXCL12/SDF-1 alpha Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CXCL12/SDF-1 alpha in cell culture supernates (10 ul), edta plasma (100 ul), platelet-poor heparin plasma (100 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 156.00 - 10000pg/mL
Assay Sensitivity 47pg/mL
Conjugate HRP
Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Platelet-poor Plasma (Heparin)
Specificity Natural and Recombinant SDF-1 alpha
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 80381
Quantity 1 Plate
Sample Volume Cell Culture Supernates (10uL),EDTA Plasma (100uL),Platelet-poor Heparin Plasma (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Target B7-H3/CD276
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.