missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCL11/I-TAC Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Brand: R&D Systems MAB672-100
This item is not returnable.
View return policy
Description
CXCL11/I-TAC Monoclonal specifically detects CXCL11/I-TAC in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| CXCL11/I-TAC | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta-R1, b-R1, chemokine (C-X-C motif) ligand 11, H174, H174IP9, Interferon gamma-inducible protein 9, Interferon-inducible T-cell alpha chemoattractant, IP-9member 11, ITAC, I-TAC, I-TACMGC102770, SCYB9B, small inducible cytokine subfamily B (Cys-X-Cys), member 9B, Small-inducible cytokine B11 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6373 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Neutralization, ELISA | |
| 87328 | |
| Western Blot 1 ug/mL, Neutralization 0.2-0.8 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| O14625 | |
| CXCL11 | |
| E. coli-derived recombinant human CXCL11/I-TAC Phe22-Phe94 Accession # O14625 | |
| 100 μg | |
| Primary | |
| Detects human CXCL11/I-TAC in ELISAs and Western blots. In ELISAs, this antibody does not cross-react with recombinant human (rh) CXCL5, 7, 8, 9, or 10. In Western blots, this antibody shows less than 1% cross-reactivity with rhCXCL1, 2, 3, 5, 6, 7, IL-8/CXCL8, 9, 10, 13, CXCL12/SDF-1 alpha , CXCL12/SDF-1 beta , rmCXCL1, 2, 6, 9, 10, CXCL12/SDF-1 alpha , rmCXCL13, rrCXCL1, CXCL3/CINC-2 alpha , CXCL3/CINC-2 beta , or rpIL-8/CXCL8. | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction