missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCL1/2/3/GRO Pan Specific Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
£180.00 - £1530.00
Specifications
| Antigen | CXCL1/2/3/GRO |
|---|---|
| Clone | 31716 |
| Dilution | Western Blot 1 ug/mL, Neutralization |
| Applications | Western Blot, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18768408
![]() |
R&D Systems
MAB276-SP |
25 μg |
£180.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18713187
![]() |
R&D Systems
MAB276-100 |
100 μg |
£387.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18723027
![]() |
R&D Systems
MAB276-500 |
500 μg |
£1530.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCL1/2/3/GRO Monoclonal specifically detects CXCL1/2/3/GRO in Human samples. It is validated for Western Blot, Neutralization.Specifications
| CXCL1/2/3/GRO | |
| Western Blot 1 ug/mL, Neutralization | |
| Monoclonal | |
| Mouse | |
| Human | |
| P19875 | |
| 2919 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 31716 | |
| Western Blot, Neutralization | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CINC1, CINC-1, CINC-2, CINC3, CINC-3, Dcip1, FSP, Gm1960, GRO alpha, GRO beta, GRO gamma, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, MGSA-beta, MIP-2, NAP-3, SCYB1 | |
| E. coli-derived recombinant human CXCL2 Ala35-Asn107 Accession # P19875 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CXCL1/2/3 in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title