missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Human CX3CL1 ELISA Kit

Product Code. 16821556 Shop All Proteintech Products
Click to view available options
Target Species:
Human
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 16821556

Brand: Proteintech™ KE0014596T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Solid phase sandwich ELISA used to detect and quantify protein levels of endogenous CX3CL1

  • Recognizes human CX3CL1
  • CXCL1 (fractalkine, FKN) is a chemokine with a transmembrane domain and chemokine domain on top of a long mucin like stalk
    • Can exist in membrane-bound form or shet soluble form, after extracellular proteolysis at membrane-proximal dibasic cleavage site
    • Soluble CX3CL1 has potent chemoattractant activity for T cells, monocytes and cell-surface-bound protein, induced by primary proinflammatory signals in activated primary endothelial cells
      • Promotes strong adhesion of leukocytes
    • Presence of CX3CL1 and receptor CX3CR1 in lymphocytes and enhanced cellular expression during inflammatory conditions suggest CX3CL1-CX3CR1 duo participates in immune-related inflammatory diseases

Process:

  • Antibody specific for CX3CL1 pre-coated onto microwells
  • CX3CL1 protein in samples captured by the coated antibody after incubation
  • After extensive washing, another horseradish peroxidase (HRP)-conjugated antibody specific for CX3CL1 is added to detect captured CX3CL1 protein
  • For signal development, followed by Tetramethyl-benzidine (TMB) reagent
  • Solution containing sulfuric acid used to stop color development and color intensity
    • Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm

Specifications

Accession Number BC001163
Assay Range 31.25 to 2000 pg/mL
Assay Sensitivity 1.9 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Human
Gene ID (Entrez) 6376
Host Species Human
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 3.5 hr.
UniProt ID P78423
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human CX3CL1 ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.