missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CUG-BP2 (aa 293-373) Control Fragment Recombinant Protein

Product Code. 30199396
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30199396

Brand: Invitrogen™ RP97001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CUG-BP1 and CUG-BP2 are proteins that bind specifically to (CUG)8 oligonucleotides in vitro. While CUG-BP1 has the major binding activity in normal cells, nuclear CUG-BP2 binding activity increases in DM cells. Both CUG-BP1 and CUG-BP2 are isoforms of a novel heterogeneous nuclear ribonucleoprotein (hnRNP), hNab50. CUG-BP1, an RNA CUG triplet repeat binding protein, regulates splicing and translation of various RNAs. Expansion of RNA CUG repeats in the DMPK in DM is associated with alterations in binding activity of CUG-BP1 as well as alterations in the translation of the C/EBPb transcription factor. CUG-BP1 is an important regulator of initiation from different AUG codons of C/EBPb mRNA.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95319
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10659
Name Human CUG-BP2 (aa 293-373) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias B230218O03; B230345P09Rik; BRUNOL3; Bruno-like protein 3; C88023; cb906; CELF 2; Celf2; CELF-2; CUG triplet repeat RNA-binding protein 2; CUG triplet repeat, RNA binding protein 2; CUG-BP- and ETR-3-like factor 2; Cugbp- and Etr3-like factor 2; CUGBP Elav-like family member 2; CUGBP, Elav-like family member 2; Cugbp2; CUG-BP2; cugbp2 protein; D230046B21Rik; elav-type ribonucleoprotein-3; elav-type RNA-binding protein 3; ETR3; ETR-3; fj87b07; hNAPOR; mETR-3; mNapor; NAPOR; napor protein isoform; napor protein long isoform; napor protein short isoform; Napor-2; neur; neuroblastoma apoptosis-related RNA-binding protein; RNA-binding protein; RNA-binding protein Brunol3; RNA-binding protein BRUNOL-3; rNapor; RP23-124P7.1; wu:fj87b07
Common Name CUG-BP2
Gene Symbol CELF2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TSTNANPLSTTSSALGALTSPVAASTPNSTAGAAMNSLTSLGTLQGLAGATVGLNNINALAGTINTPRSKRLLLPKDNNGM
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.