missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CTX-II ELISA Kit

Product Code. 18057464
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 18057464

Brand: Invitrogen™ EEL037

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

CTX-II is a biomarker for type II collagen degradation, which is mainly found in cartilage. It is a small peptide fragment that is released into the synovial fluid and blood when there is cartilage degradation. CTX-II has been extensively studied as a diagnostic and prognostic marker for osteoarthritis, and it has also been shown to be a useful tool for monitoring the efficacy of treatments targeting cartilage degradation. The structure of CTX-II is a small peptide consisting of 17 amino acids. It is derived from the C-terminal telopeptide of type II collagen. CTX-II functions as a biomarker for cartilage degradation and can be used to monitor the progression of osteoarthritis and the effectiveness of treatments.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.1 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV <10%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 100 μL, Serum 100 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target CTX-II
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 30 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.