missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CTRP9/C1qTNF9 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6537-SP
This item is not returnable.
View return policy
Description
CTRP9/C1qTNF9 Polyclonal specifically detects CTRP9/C1qTNF9 in Human samples. It is validated for Western Blot.
Specifications
| CTRP9/C1qTNF9 | |
| Polyclonal | |
| Western Blot 2 ug/mL | |
| P0C862 | |
| Sheep | |
| Antigen Affinity-purified | |
| RUO | |
| 338872 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| AQL1, C1q and tumor necrosis factor related protein 9, C1qTNF9, C1QTNF9AComplement C1q tumor necrosis factor-related protein 9, complement C1q tumor necrosis factor-related protein 9A, CTRP9, MGC48915 | |
| Mouse myeloma cell line NS0-derived recombinant human CTRP9/C1qTNF9 Asn16-Pro333 Accession # P0C862 | |
| 25 μg | |
| Primary | |
| Detects human CTRP9/C1qTNF9 in direct ELISAs. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction