missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CTRP2 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
CTRP2 Synthetic Peptide, PEP-0268, from Invitrogen.
CTRP2, also known as adiponectin C1q and collagen domain-containing protein 2, is a protein that belongs to the C1q/TNF-related protein family. It is expressed in various tissues, including adipose tissue, skeletal muscle, and the heart, and is involved in the regulation of energy metabolism and insulin sensitivity. CTRP2 has been shown to have anti-inflammatory and cardioprotective effects.
Specifications
Specifications
| Gene ID (Entrez) | 114898 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q9BXJ5 |
| Common Name | Human CTRP2 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction