missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CTGF/CCN2 C-Terminus Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF660
This item is not returnable.
View return policy
Description
CTGF/CCN2 Polyclonal specifically detects CTGF/CCN2 in Human samples. It is validated for Immunohistochemistry.
Specifications
| CTGF/CCN2 | |
| Polyclonal | |
| Unconjugated | |
| P29279 | |
| CTGF | |
| E. coli-derived recombinant human CTGF/CCN2 C-Terminus Glu247-Ala349 Accession # P29279 | |
| 100 μg | |
| Primary | |
| Detects human CTGF/CCN2 C-Terminus in direct ELISAs. In direct ELISAs, approximately 15% cross-reactivity with recombinant rat CTGF is observed. | |
| Human | |
| IgG |
| Immunohistochemistry | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL | |
| CCN2, CCN2IGFBP-8, connective tissue growth factor, CTGRP, Fisp12, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP-8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1490 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction