missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CT47 (aa 246-273) Control Fragment Recombinant Protein

Product Code. 30204535
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30204535

Brand: Invitrogen™ RP104338

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (54%), Rat (54%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59991 (PA5-59991. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CT47 (cancer/testis antigen family 47) is a 288 amino acid protein that is strongly expressed in testis and lowly expressed in placenta and in brain. The gene encoding CT47 is located on human chromosome X, which consists of about 153 million base pairs and nearly 1,000 genes. The combination of an X and Y chromosome lead to normal male development while two copies of X lead to normal female development. There are a number of conditions related to an unsual number and combination of sex chromosomes being inherited. More than one copy of the X chromosome with a Y chromosome causes Klinefelter's syndrome. A single copy of X alone leads to Turner's syndrome. More than 2 copies of the X chromosome, in the absence of a Y chromosome, is known as Triple X syndrome. Color blindness, hemophilia, and Duchenne muscular dystrophy are well known X chromosome-linked conditions which affect males more frequently as males carry a single X chromosome.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5JQC4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 728096
Name Human CT47 (aa 246-273) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Cancer/testis antigen 47; Cancer/testis antigen 47 A; cancer/testis antigen family 47, member A1; cancer/testis CT47 family, member 1; CT47; CT47.1; CT47.10; CT47.11; CT47.12; CT47.2; CT47.3; CT47.4; CT47.5; CT47.6; CT47.7; CT47.8; CT47.9; CT47A; CT47A1; CT47A10; CT47A11; CT47A12; CT47A2; CT47A3; CT47A4; CT47A5; CT47A6; CT47A7; CT47A8; CT47A9
Common Name CT47
Gene Symbol CT47A1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PEEVTKSQPEKWDEEAQDAAGEEEKEQE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.