missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CSRP2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF6196
This item is not returnable.
View return policy
Description
CSRP2 Polyclonal specifically detects CSRP2 in Human samples. It is validated for Western Blot.
Specifications
| CSRP2 | |
| Polyclonal | |
| Unconjugated | |
| Q16527 | |
| CSRP2 | |
| E. coli-derived recombinant human CSRP2 Met1-Gln193 Accession # Q16527 | |
| 100 μg | |
| Primary | |
| Detects human CSRP2 in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| CRP2, CRP2LIM domain only protein 5, cysteine and glycine-rich protein 2, Cysteine-rich protein 2, LMO5LMO-5, SMLIM, SmLIMLIM domain only 5, smooth muscle, Smooth muscle cell LIM protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1466 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction