missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CSGalNAcT-2 (NP_061060) VersaClone cDNA

Product Code. 18796538
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18796538

Brand: R&D Systems™ RDC1351

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

The Human CSGalNAcT-2 (NP_061060) VersaClone cDNA clone contains the complete ORF for CSGALNACT2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CSGALNACT2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature

TRUSTED_SUSTAINABILITY

Specifications

Gene CSGALNACT2
Accession Number NP_061060
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.24
Insert Type 1641bp
Gene ID (Entrez) 55454
Gene Alias beta4GalNAcT-2, CHGN2, chondroitin beta1,4 N-acetylgalactosaminyltransferase 2, Chondroitin beta-1,4-N-acetylgalactosaminyltransferase 2, chondroitin sulfate GalNAcT-2, chondroitin sulfate N-acetylgalactosaminyltransferase 2, CSGalNAcT2, DKFZp686H13226, EC 2.4.1.174, FLJ43310, GALNACT2, GALNACT-2, GALNACT2beta 4 GalNAcT-2, MGC40204, PRO0082
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg

For Reasearch Use Only. Not for use in Humans.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.