missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CRYZL1 (aa 73-160) Control Fragment Recombinant Protein

Product Code. 30196164
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30196164

Brand: Invitrogen™ RP95906

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56131 (PA5-56131. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a protein that has sequence similarity to zeta crystallin, also known as quinone oxidoreductase. This zeta crystallin-like protein also contains an NAD(P)H binding site. Alternatively spliced transcript variants have been observed but their full-length nature has not been completely determined.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95825
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9946
Name Human CRYZL1 (aa 73-160) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2210407J23Rik; 2410006O11Rik; 4P11; crystallin zeta like 1; crystallin, zeta (quinone reductase)-like 1; crystallin, zeta like 1; crystallin, zeta-like 1 isoform a; Cryzl1; dZ63M10.5; protein 4P11; QOH-1; quinone oxidoreductase homolog 1; Quinone oxidoreductase-like protein 1; quinone oxidoreductase-like protein 1; LOW QUALITY PROTEIN: quinone oxidoreductase-like protein 1; quinone reductase-like 1; zeta-crystallin homolog; zgc:92225
Common Name CRYZL1
Gene Symbol CRYZL1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VGSKVSFFQPDDEVVGILPLDSEDPGLCEVVRVHEHYLVHKPEKVTWTEAAGSIRDGVRAYTALHYLSHLSPGKSVLIMDGASAFGTI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.