missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cryptic Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF1410
This item is not returnable.
View return policy
Description
CYRPTIC/CFC1 Polyclonal specifically detects CYRPTIC/CFC1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| CYRPTIC/CFC1 | |
| Polyclonal | |
| Unconjugated | |
| NP_115934 | |
| CFC1 | |
| Mouse myeloma cell line NS0-derived recombinant human Cryptic Tyr26-Gly169 Accession # NP_115934 | |
| 100 μg | |
| Primary | |
| Detects human Cryptic in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse Cryptic is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| CFC1, CFC-1, CFC1B, cripto, FRL-1, cryptic family 1, Cryptic family protein 1, cryptic protein, CRYPTICFLJ77897, HTX2MGC133213 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 55997 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction