missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CRTAM Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB16951
This item is not returnable.
View return policy
Description
CRTAM Monoclonal specifically detects CRTAM in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| CRTAM | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| O95727 | |
| CRTAM | |
| Mouse myeloma cell line NS0-derived recombinant human CRTAM Ser18-Ser286 Accession # O95727 | |
| 500 μg | |
| Primary | |
| Detects human CRTAM in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CRTAM is observed. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 210213 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD355, CD355 antigen, class I MHC restricted T cell associated molecule, class-I MHC-restricted T cell associated molecule, Class-I MHC-restricted T-cell-associated molecule, cytotoxic and regulatory T cell molecule, cytotoxic and regulatory T-cell molecule | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 56253 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction