missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CRP2 (aa 98-122) Control Fragment Recombinant Protein

Código de producto. 30180945
Click to view available options
Quantity:
100 μL
Tamaño de la unidad:
100µL
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30180945

Marca: Invitrogen™ RP99041

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83785 (PA5-83785. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CSRP2 is a member of the CSRP family of genes, encoding a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. CRP2 contains two copies of the cysteine-rich amino acid sequence motif (LIM) with putative zinc-binding activity, and may be involved in regulating ordered cell growth. Other genes in the family include CSRP1 and CSRP3.
TRUSTED_SUSTAINABILITY

Especificaciones

Accession Number Q16527
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 1466
Name Human CRP2 (aa 98-122) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AW551867; CRP2; CSRP2; cysteine and glycine rich protein 2; cysteine and glycine-rich protein 2; cysteine rich protein 2; cysteine-rich protein 2; Dlp1; DLP-1; Double LIM protein 1; LIM domain only 5, smooth muscle; LIM domain only protein 5; LIM/double zinc-finger protein; LMO5; LMO-5; SmLIM; Smooth muscle cell LIM protein
Common Name CRP2
Gene Symbol CSRP2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence HRPTTNPNTSKFAQKYGGAEKCSRC
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado