missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CRMP5 (aa 241-326) Control Fragment Recombinant Protein

Product Code. 30211958
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30211958

Brand: Invitrogen™ RP105561

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84896 (PA5-84896. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The collapsin response mediator protein (CRMP) family of proteins (also referred to as ULIP6) are a group of neurologic proteins that have been found to play key roles in growth cone guidance during neural development. CRMP5 has relatively low sequence homology with the other four members of the CRMP family. CRMP5 is expressed in the developing nervous system in an expression pattern that resembles CRMP2 and has a peak expression during the first postnatal week. Elevated levels of autoantibody to CRMP5 have been shown to be linked to paraneoplastic autoimmune optic neuritis and other paraneoplastic neoplasms. Auto-immunity against CRMP5 expression (predominantly the N-terminal epitopes) is also being investigated as an oncological marker in the respiratory system and in the thymus.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BPU6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 56896
Name Human CRMP5 (aa 241-326) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias collapsin response mediator protein 5; collapsin response mediator protein-5; CRAM; CRMP3-associated molecule; Crmp5; CRMP-5; dihydropyrimidinase like 5; dihydropyrimidinase-like 5; dihydropyrimidinase-related protein 5; Dpysl5; DRP-5; Ulip6; ULIP-6; ULIP6 protein; UNC33-like phosphoprotein 6
Common Name CRMP5
Gene Symbol DPYSL5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence CPIYLVNVSSISAGDVIAAAKMQGKVVLAETTTAHATLTGLHYYHQDWSHAAAYVTVPPLRLDTNTSTYLMSLLANDTLNIVASDH
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.