missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CRLR Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10044-SP
This item is not returnable.
View return policy
Description
CRLR Monoclonal specifically detects CRLR in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Specifications
| CRLR | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| calcitonin gene-related peptide type 1 receptor, calcitonin receptor-like, CALCRL, CGRP type 1 receptor, CGRPR, CGRPRCRLRCalcitonin receptor-like receptor | |
| Synthetic peptide containing human CALCRL, Accession No. Q16602 | |
| 25 μg | |
| 10203.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
| Immunohistochemistry, Immunocytochemistry | |
| 998820 | |
| Immunohistochemistry 5-25 μg/mL, Immunocytochemistry 8-25 μg/mL | |
| Q16602 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human CALCRL in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction