missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human CRISP-2 - Ready-To-Use ELISA Kit (Colorimetric)

Product Code. 30057008
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30057008 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30057008 Supplier Novus Biologicals™ Supplier No. NBP331320

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Colorimetric ELISA Kit

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.156 - 10 ng/mL (example only; lot dependent)
Assay Sensitivity 0.055 ng/mL (example only; lot dependent)
Product Type ELISA Kit (Colorimetric)
Sample Type Tissue Homogenates, Biological Fluids
For Use With (Application) ELISA
Gene ID (Entrez) 7180
Gene Symbol CRISP2
Quantity 1 Kit
Research Discipline Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym Cancer/testis antigen 36, CRISP-2GAPDL5, CT36TSP1, cysteine-rich secretory protein 2, glyceraldehyde-3-phosphate dehydrogenase-like 5, testis specific protein 1 (probe H4-1 p3-1), Testis-specific protein TPX-1, TPX1MGC111136
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.