missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CRISP-2 ELISA Kit

Product Code. 17814070
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
This item is not returnable. View return policy

Product Code. 17814070

Brand: Invitrogen™ EH130RBX10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human CRISP-2 quantitates human CRISP-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CRISP-2.

The cysteine-rich secretory proteins (CRISP) family is a group of four proteins that are strongly expressed in the male reproductive tract and have been implicated in having roles in male fertility. CRISP2, also known as TPX1, has been implicated in the adhesion between spermatids and Sertoli cells, and with CRISP1, is thought to be involved in sperm-egg fusion. CRISP2 has been shown to regulate the Ca2+ influx through ryanodine receptors (RYR) and may influence the acrosome reaction or sperm motility. CRISP2 has also been shown to bind to the mitogen-activated protein kinase kinase kinase 11 (MAP3K11) and localizes to the developing acrosome, suggesting this CRISP2-MAP3K11 complex may have a role in acrosome development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Cancer/testis antigen 36; Crisp2; CRISP-2; Crisp2l; CT36; cysteine rich secretory protein 2; cysteine-rich secretory protein 2; cysteine-rich secretory protein 2-like; GAPDL5; glyceraldehyde-3-phosphate dehydrogenase-like 5; MGC111136; OTTHUMP00000016589; testicular tissue protein Li 43; testis specific gene 1; testis specific protein 1; testis specific protein 1 (probe H4-1 p3-1); testis-specific protein TPX-1; TPX1; Tpx-1; TSP1
Assay Range 0.65 to 150 ng/mL
Assay Sensitivity 0.65 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 7180
Gene Symbol CRISP2
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target CRISP2
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.