missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CRISP-2 ELISA Kit
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
Description
Human CRISP-2 quantitates human CRISP-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CRISP-2.
The cysteine-rich secretory proteins (CRISP) family is a group of four proteins that are strongly expressed in the male reproductive tract and have been implicated in having roles in male fertility. CRISP2, also known as TPX1, has been implicated in the adhesion between spermatids and Sertoli cells, and with CRISP1, is thought to be involved in sperm-egg fusion. CRISP2 has been shown to regulate the Ca2+ influx through ryanodine receptors (RYR) and may influence the acrosome reaction or sperm motility. CRISP2 has also been shown to bind to the mitogen-activated protein kinase kinase kinase 11 (MAP3K11) and localizes to the developing acrosome, suggesting this CRISP2-MAP3K11 complex may have a role in acrosome development.
Specifications
Specifications
| Accession Number | Cancer/testis antigen 36; Crisp2; CRISP-2; Crisp2l; CT36; cysteine rich secretory protein 2; cysteine-rich secretory protein 2; cysteine-rich secretory protein 2-like; GAPDL5; glyceraldehyde-3-phosphate dehydrogenase-like 5; MGC111136; OTTHUMP00000016589; testicular tissue protein Li 43; testis specific gene 1; testis specific protein 1; testis specific protein 1 (probe H4-1 p3-1); testis-specific protein TPX-1; TPX1; Tpx-1; TSP1 |
| Assay Range | 0.65 to 150 ng/mL |
| Assay Sensitivity | 0.65 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 7180 |
| Gene Symbol | CRISP2 |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction