missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CRISP-2 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF2575
This item is not returnable.
View return policy
Description
CRISP-2 Polyclonal specifically detects CRISP-2 in Human samples. It is validated for Western Blot.
Specifications
| CRISP-2 | |
| Polyclonal | |
| Biotin | |
| P16562 | |
| CRISP2 | |
| Mouse myeloma cell line NS0-derived recombinant human CRISP-2 Lys22-Tyr243 Accession # P16562 | |
| 50 μg | |
| Primary | |
| Detects human CRISP-2 in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human CRISP-3 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Cancer/testis antigen 36, CRISP2, CRISP-2GAPDL5, CT36TSP1, cysteine-rich secretory protein 2, GAPDL5, glyceraldehyde-3-phosphate dehydrogenase-like 5, testis specific protein 1 (probe H4-1 p3-1), Testis-specific protein TPX-1, TPX1, TPX1MGC111136, TSP1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7180 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction