missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CRISP-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2575
This item is not returnable.
View return policy
Description
CRISP-2 Monoclonal specifically detects CRISP-2 in Human samples. It is validated for Western Blot.
Specifications
| CRISP-2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P16562 | |
| CRISP2 | |
| Mouse myeloma cell line NS0-derived recombinant human CRISP-2 Lys22-Tyr243 Accession # P16562 | |
| 100 μg | |
| Primary | |
| Detects human CRISP-2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human CRISP-3 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 350814 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cancer/testis antigen 36, CRISP2, CRISP-2GAPDL5, CT36TSP1, cysteine-rich secretory protein 2, GAPDL5, glyceraldehyde-3-phosphate dehydrogenase-like 5, testis specific protein 1 (probe H4-1 p3-1), Testis-specific protein TPX-1, TPX1, TPX1MGC111136, TSP1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7180 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction