missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CRIM1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1917-SP
This item is not returnable.
View return policy
Description
CRIM1 Monoclonal specifically detects CRIM1 in Human samples. It is validated for Western Blot.
Specifications
| CRIM1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9NZV1 | |
| CRIM1 | |
| Mouse myeloma cell line NS0-derived recombinant human CRIM1 Leu35-Ser939 Accession # Q9NZV1 | |
| 25 μg | |
| Primary | |
| Detects human CRIM1 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 253511 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CRIM-1, cysteine rich transmembrane BMP regulator 1 (chordin-like), cysteine-rich motor neuron 1, cysteine-rich motor neuron 1 protein, Cysteine-rich repeat-containing protein S52, MGC138194, S52 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 51232 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction