missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CRHBP Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF2796-SP
This item is not returnable.
View return policy
Description
CRHBP Polyclonal specifically detects CRHBP in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CRHBP | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| corticotropin releasing hormone binding protein, corticotropin-releasing factor-binding protein, Corticotropin-releasing hormone-binding protein, CRFBP, CRFBPcorticotropin releasing hormone-binding protein, CRF-BPCRF-binding protein, CRH-BP | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1393 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P24387.2 | |
| CRHBP | |
| Mouse myeloma cell line NS0-derived recombinant human CRHBP Tyr25-Leu322 Accession # P24387.2 | |
| 25 μg | |
| Primary | |
| Detects human CRHBP in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant mouse CRHBP is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction