missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CRELD2 alpha Isoform Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3555
Additional Details : Weight : 0.00970kg
Description
CRELD2 Polyclonal specifically detects CRELD2 in Human samples. It is validated for Western Blot.Specifications
CRELD2 | |
Polyclonal | |
Unconjugated | |
AAH50675 | |
CRELD2 | |
Mouse myeloma cell line NS0-derived recombinant human CRELD2 alpha Isoform Ala21-Leu353 Accession # AAH50675 | |
100 μg | |
Primary | |
Detects human CRELD2 alpha isoform in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant mouse (rm) CRELD2 is observed and less than 1% cross-reactivity with recombinant human CRELD1 and rmCRELD1 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
cysteine-rich with EGF-like domain protein 2, cysteine-rich with EGF-like domains 2, DKFZp667O055, MGC11256 | |
Goat | |
Affinity Purified | |
RUO | |
79174 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only