missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CRELD1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3128
Additional Details : Weight : 0.00970kg
Description
CRELD1 Polyclonal specifically detects CRELD1 in Human samples. It is validated for Western Blot.Specifications
CRELD1 | |
Polyclonal | |
Unconjugated | |
Q96HD1 | |
CRELD1 | |
Mouse myeloma cell line NS0-derived recombinant human CRELD1 isoform 1 Gln30-Glu362 Accession # Q96HD1 | |
100 μg | |
Primary | |
Detects human CRELD1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
atrioventricular septal defect 2, AVSD2, CIRRIN, cysteine-rich with EGF-like domain protein 1, cysteine-rich with EGF-like domains 1, DKFZp566D213 | |
Goat | |
Affinity Purified | |
RUO | |
78987 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only