missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CREG Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2380-SP
This item is not returnable.
View return policy
Description
CREG Monoclonal specifically detects CREG in Human samples. It is validated for Western Blot.
Specifications
| CREG | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O75629 | |
| CREG1 | |
| Mouse myeloma cell line NS0-derived recombinant human CREG Arg32-Gln220 Accession # O75629 | |
| 25 μg | |
| Primary | |
| Detects human CREG in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CREG is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 299133 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cellular repressor of E1A-stimulated genes, cellular repressor of E1A-stimulated genes 1CREG, CREG1, protein CREG1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8804 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction