missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CREBZF Synthetic Peptide

Product Code. 15275219
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 15275219

Brand: Invitrogen™ PEP1220

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

ZHANGFEI Synthetic Peptide, PEP-1220, from Invitrogen.

Strongly activates transcription when bound to HCFC1. Suppresses the expression of HSV proteins in cells infected with the virus in a HCFC1-dependent manner. Also suppresses the HCFC1-dependent transcriptional activation by CREB3 and reduces the amount of CREB3 in the cell. Able to down-regulate expression of some cellular genes in CREBZF-expressing cells.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 58487
Species Human
Purification Method purified
Accession Number Q9NS37
Common Name Human CREBZF
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol CREBZF
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.