missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CREB3L3 (aa 125-214) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (56%), Rat (56%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59244 (PA5-59244. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
CREB3L3 is a liver-specific transcription factor and member of the CREB/ATF transcription factor family (Omori et al., 2001 [PubMed 11353085]).
Specifications
Specifications
| Accession Number | Q68CJ9 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 84699 |
| Name | Human CREB3L3 (aa 125-214) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | BC010786; cAMP responsive element binding protein 3 like 3; cAMP responsive element binding protein 3-like 3; cAMP-responsive element-binding protein 3-like protein 3; CREB/ATF family transcription factor; CREB3L3; CREBH; CREB-H; Cyclic AMP-responsive element-binding protein 3-like protein 3; D10Bur1e; HYST1481; Processed cyclic AMP-responsive element-binding protein 3-like protein 3; Transcription factor CREB-H |
| Common Name | CREB3L3 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction