missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CPEB1 (aa 545-562) Synthetic Peptide
Description
Human CPEB (residues 545-562) Synthetic Peptide, PEP-171, from Invitrogen.
PEP-171 is an 18 amino acid synthetic peptide whose sequences are derived from human CPEB protein. The sequence of this peptide is (amino to carboxy terminus): C-H (545)-S-M-E-G-L-R-H-H-S-P-L-M-R-N-Q-K-N (562)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and the human CPEB, catalog # PA1-1100. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.
Specifications
Specifications
| Gene ID (Entrez) | 64506 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q9BZB8 |
| Common Name | Human CPEB1 (aa 545-562) |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Lyophilized |
| Formulation | peptide with no preservative |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?